Anti-CD86 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: GL-1]
Catalog # ROCK200-502-N90
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:CD86 molecule
- Antigen symbol:CD86
- Clonality:Monoclonal
- Clone:GL-1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:12524
- Antigen synonyms:CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
- Storage temperature:Cool
- Concentration:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-CD86 Antibody (Monoclonal) was produced by repeated immunizations with CD86 antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Anti-CD86 is useful for immunochemistry, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze! Light sensitive!
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse