Anti-RAD9 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-468
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:RAD9 homolog A (S. pombe)
- Antigen symbol:RAD9A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Yeast,S. cerevisiae
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:603761
- Amino acid number:1249 to 1263
- Modification:phospho-Ser1260
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,41 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to phosphorylated form of aa 1249-1263 of 1309 of yeast Rad9 Protein conjugated to KLH.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This phospho specific polyclonal antibody was tested by immunoblotting and ELISA. Data from both immunoblotting and ELISA indicate the antibody is reactive with the phosphorylated form of the immunizing peptide and minimally reactive with the non-phospho
Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:500- 1:2000;
Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae, Yeast