Order Entry
Denmark
ContactUsLinkComponent
 
Anti-COL2 Mouse Monoclonal Antibody [clone: II-4C11]
  CP18-100
Anti-COL2 Mouse Monoclonal Antibody [clone: II-4C11]
  CP18-100

 

  • Antibody type:
    Primary
  • Antigen name:
    collagen, type II
  • Antigen symbol:
    COL2
  • Clonality:
    Monoclonal
  • Clone:
    II-4C11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: monoclonal
Clone: II-4C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: