Anti-SEPARIN Rabbit Polyclonal Antibody
Catalog # PIEAPA512691
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen symbol:Separase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:9700
- Antigen synonyms:Separin|Caspase-like protein ESPL1|Separase|KIAA0165|ESP1|Extra spindle poles-like 1 protein|ESPL1
- Modification:Phosphorylated: Ser1126
- Storage buffer:PBS, 0.09% sodium azide
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S1126 of human SEPARIN
- Purification:Antigen affinity chromatography
- Size:400 µL
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SEPARIN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human