Anti-Tropomyosin Mouse Monoclonal Antibody [clone: TM-33]
BSBTMA1095
- Antibody type:Primary
- Antigen name:Tropomyosin 1
- Antigen symbol:TPM1
- Clonality:Monoclonal
- Clone:TM-33
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:alpha tropomyosin|TMSA|hypertrophic 3|alpha-tropomyosin|sarcomeric tropomyosin kappa|C15orf13|tropomyosin alpha-1 chain|tropomyosin-1|cardiomyopathy|CMD1Y|CMH3
- Molecular weight:36/39 kDa
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Chicken gizzard tropomyosin.
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for Tropomyosin (36/39 kDa), tropomyosin 1 (alpha) (TPM1) detection. Tested with WB in Human; mouse; rat; chicken. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Tropomyosin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken