Anti-FPR2 Rabbit Polyclonal Antibody
The FPRL1 / FPR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FPRL1 / FPR2. This antibody reacts with human, mouse, rat. The FPRL1 / FPR2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:300
Type: Primary
Antigen: FPRL1/FPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- NOVUNLS1878
- NOVUNLS1878SS
- Antigen symbol:
- FPRL1
- FPRL1
- Antigen name:
- Formyl Peptide Receptor-like 1
- Formyl Peptide Receptor-like 1
- Antigen synonyms:
- FPRH1|fMLPR1|HM63|LXA4R|formyl peptide Receptor 2|ALXR
- FPRH1|fMLPR1|HM63|LXA4R|formyl peptide Receptor 2|ALXR
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2358
- 2358
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together