Order Entry
Denmark
ContactUsLinkComponent
Anti-FHOD3 Rabbit Polyclonal Antibody
Anti-FHOD3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24544
Supplier:  Novus Biologicals
undefined
Anti-FHOD3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24544
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Formin homology 2 domain containing 3
  • Antigen symbol:
    FHOD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Gene ID:
    80206
  • Antigen synonyms:
    FHOS2|Formactin2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: FHOD3 (formin homology 2 domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: