Anti-PACS1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24496
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:phosphofurin acidic cluster sorting protein 1
- Antigen symbol:PACS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:55690
- Antigen synonyms:MRD17
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: PACS1 (phosphofurin acidic cluster sorting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: