Anti-ERManI Mouse Monoclonal Antibody [clone: 3C2]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ERManI
Clonality: monoclonal
Clone: 3C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP2-13167
- NOVUNBP2-13167SS
- Antigen symbol:
- MAN1B1
- MAN1B1
- Antigen name:
- Mannosidase alpha class 1B member 1
- Mannosidase alpha class 1B member 1
- Antigen synonyms:
- MRT15|MANA-ER|ERMAN1
- MRT15|MANA-ER|ERMAN1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 3C2
- 3C2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 11253
- 11253
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together