Order Entry
Denmark
ContactUsLinkComponent
Anti-LC3B Rabbit Polyclonal Antibody
Anti-LC3B Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-LC3B Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB600-1384
  • NOVUNB600-1384SS
  • Antigen symbol:
  • LC3B
  • LC3B
  • Antigen name:
  • Microtubule-associated Protein 1 light chain 3 beta
  • Microtubule-associated Protein 1 light chain 3 beta
  • Antigen synonyms:
  • MAP1LC3B|Autophagy-related protein LC3 B|MAP1 light chain 3-like protein 2|MAP1ALC3|MAP1A/MAP1B LC3 B|Microtubule-associated proteins 1A/1B light chain 3B [Precursor]|MAP1A/1B light chain 3 B|Autophagy-related ubiquitin-like modifier LC3 B
  • MAP1LC3B|Autophagy-related protein LC3 B|MAP1 light chain 3-like protein 2|MAP1ALC3|MAP1A/MAP1B LC3 B|Microtubule-associated proteins 1A/1B light chain 3B [Precursor]|MAP1A/1B light chain 3 B|Autophagy-related ubiquitin-like modifier LC3 B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 81631
  • 81631
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together