Order Entry
Denmark
ContactUsLinkComponent
Anti-Aquaporin 4 Rabbit Polyclonal Antibody
Anti-Aquaporin 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1937
Supplier:  Boster Bio
undefined
Anti-Aquaporin 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1937
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Aquaporin 4
  • Antigen symbol:
    AQP4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    WCH4|AQP-4|Aquaporin4|MIWC|Aquaporin-4|Mercurial-insensitive water channel
  • Amino acid number:
    258 to 274
  • UniProtKB:
    P55087
  • Sequence:
    FKRRFKEAFSKAAQQTK
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 4 (258-274aa FKRRFKEAFSKAAQQTK), different from the related rat and mouse sequences by two amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Aquaporin-4(AQP4) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

AQP4(Aquaporin 4), also known as MERCURIAL-INSENSITIVE WATER CHANNEL; MIWC, is protein which in humans is encoded by the AQP4 gene. The aquaporins are a family of water-selective membrane channels found in animals, plants, and microorganisms. AQP4 is the predominant water channel in the brain and has an important role in brain water homeostasis. By fluorescence in situ hybridization, Lu et al.(1996) determined that the AQP4 gene maps to 18q11.2-q12.1. By interspecific backcross analysis, Turtzo et al.(1997) mapped the mouse Aqp4 gene to the proximal region of chromosome 18. Analyzing the expression of AQP4 in mammalian skeletal muscle, Frigeri et al.(1998) found that, in immunohistochemical experiments, affinity-purified AQP4 antibodies stained selectively the sarcolemma of fast-twitch fibers. Immunocytochemistry revealed strong AQP4 water channel expression in Muller cells in mouse retina and in fibrous astrocytes in optic nerve.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

WB: The detection limit for AQP4 is approximately 0.5ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: Aquaporin 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat