Specifications
- Antibody type:Primary
- Antigen name:cholinergic receptor, nicotinic, alpha 1 (muscle)
- Antigen symbol:CHRNA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Lyophilized
- Antigen synonyms:CMS2A|SCCMS|CMS1B|ACHRD|ACHRA|CHRNA|FCCMS|CMS1A
- UniProtKB:P02708
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Nicotinic Acetylcholine Receptor alpha 1(22-36aa EHETRLVAKLFKDYS), identical to the related rat sequence, different from the related mouse sequence by one amino acid.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Acetylcholine receptor subunit alpha(CHRNA1) detection. Tested with WB in Human;Mouse;Rat.
CHRNA, also termed ACHRA, is mapped on 2q24-q32.This gene encodes the alpha subunit of the muscle acetylcholine receptor, which is the main target of pathogenic autoantibodies in autoimmune myasthenia gravis. The protein-coding sequence of the human alpha subu gene is divided into 9 exons that correspond to different structural and functional domains of the precursor molecule.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: CHRNA1 (cholinergic Receptor, nicotinic, alpha 1 (muscle))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat