Order Entry
Denmark
ContactUsLinkComponent
Anti-Collagen, Type IV Mouse Monoclonal Antibody [clone: Col-30]
Anti-Collagen, Type IV Mouse Monoclonal Antibody [clone: Col-30]
Catalog # BSBTMA1030
Supplier:  Boster Bio
undefined
Anti-Collagen, Type IV Mouse Monoclonal Antibody [clone: Col-30]
Catalog # BSBTMA1030
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collagen, Type IV
  • Antigen symbol:
    COL4
  • Clonality:
    Monoclonal
  • Clone:
    Col-30
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Human collagen type IV
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for Collagen, Type IV, collagen, type IV, alpha-1; collagen, type IV, alpha 3 (Goodpasture antigen); collagen,type IV, alpha 4; collagen, type IV, alpha 5; collagen, type IV, alpha 6 (COL4A1; COL4A3; COL4A4; COL4A5; COL4A6) detection. Tested with IHC-P, IHC-F in Human. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Collagen, Type IV
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human