Anti-NLRP12 Mouse Monoclonal Antibody [clone: 3B5]
Catalog # ABNOH00091662-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NACHT, LRR And PYD Domains-Containing protein 12
- Antigen symbol:NLRP12
- Clonality:Monoclonal
- Clone:3B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:91662
- Antigen synonyms:PAN6|RNO2|RNO|NALP12|PYPAF7|FCAS2|CLR19.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NLRP12
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human