Anti-DPYSL5 Mouse Monoclonal Antibody [clone: 2D10]
ABNOH00056896-M04
: Abnova
- Antibody type:Primary
- Antigen name:Dihydropyrimidinase-like 5
- Antigen symbol:DPYSL5
- Clonality:Monoclonal
- Clone:2D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:56896
- Antigen synonyms:CRMP-5|CRMP5|CRAM|Ulip6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: DPYSL5
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat