Anti-RETN Rabbit Polyclonal Antibody
ABNOH00056729-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Resistin
- Antigen symbol:RETN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:56729
- Antigen synonyms:Cysteine-rich secreted protein FIZZ3|Adipose tissue-specific secretory factor|C/EBP-epsilon-regulated myeloid-specific secreted cysteine-|RSN|ADSF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human