Anti-SSH1 Mouse Monoclonal Antibody [clone: 1D2]
ABNOH00054434-M02
: Abnova
- Antibody type:Primary
- Antigen name:Slingshot Protein Phosphatase 1
- Antigen symbol:SSH1
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:54434
- Antigen synonyms:SSH1L
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SSH1
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human