Anti-ARMCX1 Mouse Monoclonal Antibody [clone: 6E10]
ABNOH00051309-M01
: Abnova
- Antibody type:Primary
- Antigen name:Armadillo Repeat Containing, X-linked 1
- Antigen symbol:ARMCX1
- Clonality:Monoclonal
- Clone:6E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51309
- Antigen synonyms:AD032|ALEX1|ARM protein lost in epithelial cancers on chromosome X 1|Armadillo repeat-containing X-linked protein 1|ARMCX1|Protein ALEX1|GASP7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ARMCX1
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human