Anti-PRSS50 Mouse Monoclonal Antibody [clone: 2D8]
ABNOH00029122-M10
: Abnova
- Antibody type:Primary
- Antigen name:Protease Serine 50
- Antigen symbol:PRSS50
- Clonality:Monoclonal
- Clone:2D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:29122
- Antigen synonyms:TSP50|CT20
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PRSS50
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human