Anti-USP21 Mouse Monoclonal Antibody [clone: 3D10]
Catalog # ABNOH00027005-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 21
- Antigen symbol:USP21
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:27005
- Antigen synonyms:USP16|USP23
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: USP21
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human