Anti-GREM1 Mouse Monoclonal Antibody [clone: 2C8]
Catalog # ABNOH00026585-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gremlin 1
- Antigen symbol:GREM1
- Clonality:Monoclonal
- Clone:2C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26585
- Antigen synonyms:Down-regulated in Mos-transformed cells protein|CKTSF1B1|DRM|DAND2|Increased in high glucose protein 2|DAN domain family member 2|Gremlin-1|Cysteine knot superfamily 1|Cell proliferation-inducing gene 2 protein|BMP antagonist 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: GREM1
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human