Anti-PSIP1 Mouse Monoclonal Antibody [clone: 1C4]
Catalog # ABNOH00011168-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:PC4 and SFRS1 interacting protein 1
- Antigen symbol:PSIP1
- Clonality:Monoclonal
- Clone:1C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:11168
- Antigen synonyms:p52|p75|LEDGF|PSIP2|DFS70|PAIP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PSIP1
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human