Anti-CCT2 Mouse Monoclonal Antibody [clone: 2G6]
ABNOH00010576-M01
: Abnova
- Antibody type:Primary
- Antigen name:Chaperonin Containing TCP1, subunit 2 (beta)
- Antigen symbol:CCT2
- Clonality:Monoclonal
- Clone:2G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10576
- Antigen synonyms:MGC142074|TCP 1 beta|MGC94480|TCP-1-beta|T complex protein 1 beta subunit|TCPB_HUMAN|Chaperonin containing TCP1 subunit 2 beta|PRO1633|CTP:phosphocholine cytidylyltransferase 2|CCTB|T-complex protein 1 subunit beta|T complex protein 1 subunit beta|MGC142076|CCT-2|99D8.1.|CCT 2|CCT-beta|Chaperonin containing t complex polypeptide 1 beta subunit|Chaperonin containing TCP1 subunit 2|CCT beta|Chaperonin containing t complex polypeptide 1 subunit 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CCT2
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human