Order Entry
Denmark
ContactUsLinkComponent
Anti-OLFM1 Mouse Monoclonal Antibody [clone: 2E9-1A2]
Anti-OLFM1 Mouse Monoclonal Antibody [clone: 2E9-1A2]
Catalog # ABNOH00010439-M02
Supplier:  Abnova
undefined
Anti-OLFM1 Mouse Monoclonal Antibody [clone: 2E9-1A2]
Catalog # ABNOH00010439-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Olfactomedin 1
  • Antigen symbol:
    OLFM1
  • Clonality:
    Monoclonal
  • Clone:
    2E9-1A2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    10439
  • Antigen synonyms:
    NOE1|OLFM1|NOEL1|Neuronal olfactomedin-related ER localized protein|Olfactomedin-1|Noelin
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: OLFM1
Clonality: Monoclonal
Clone: 2E9-1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human