Order Entry
Denmark
ContactUsLinkComponent
 
Anti-FAM65B Mouse Monoclonal Antibody [clone: 2E6-1B10]
Catalog # ABNOH00009750-M01
Supplier:  Abnova
undefined
Anti-FAM65B Mouse Monoclonal Antibody [clone: 2E6-1B10]
Catalog # ABNOH00009750-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 65, member B
  • Antigen symbol:
    FAM65B
  • Clonality:
    Monoclonal
  • Clone:
    2E6-1B10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    9750
  • Antigen synonyms:
    DIFF40|MYONAP|PL48|C6orf32|DIFF48
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: FAM65B
Clonality: Monoclonal
Clone: 2E6-1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human