Anti-TM9SF2 Mouse Monoclonal Antibody [clone: 1C2]
ABNOH00009375-M12
: Abnova
- Antibody type:Primary
- Antigen name:Transmembrane 9 Superfamily Member 2
- Antigen symbol:TM9SF2
- Clonality:Monoclonal
- Clone:1C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9375
- Antigen synonyms:P76
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TM9SF2
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human