Anti-HERC3 Mouse Monoclonal Antibody [clone: 3G7]
ABNOH00008916-M02
: Abnova
- Antibody type:Primary
- Antigen name:HECT and RLD domain containing E3 ubiquitin protein ligase 3
- Antigen symbol:HERC3
- Clonality:Monoclonal
- Clone:3G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8916
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:100 µg
Type: Primary
Antigen: HERC3
Clonality: Monoclonal
Clone: 3G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human