Anti-TNKS Mouse Monoclonal Antibody [clone: 1A3]
ABNOH00008658-M02
: Abnova
- Antibody type:Primary
- Antigen name:Tankyrase
- Antigen symbol:TNKS
- Clonality:Monoclonal
- Clone:1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8658
- Antigen synonyms:PARP-5a|TINF1|TNKS1|TIN1|PARP5A|pART5|PARPL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TNKS
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human