Anti-BBOX1 Mouse Monoclonal Antibody [clone: 6H3]
Catalog # ABNOH00008424-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:2-Oxoglutarate Dioxygenase 1
- Antigen symbol:BBOX1
- Clonality:Monoclonal
- Clone:6H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8424
- Antigen synonyms:BBOX|gamma-BBH|BBH|G-BBH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BBOX1
Clonality: Monoclonal
Clone: 6H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human