Anti-ZIC3 Mouse Monoclonal Antibody [clone: 3C6]
ABNOH00007547-M07
: Abnova
- Antibody type:Primary
- Antigen name:Zinc Finger Protein of the cerebellum 3
- Antigen symbol:ZIC3
- Clonality:Monoclonal
- Clone:3C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7547
- Antigen synonyms:HTX|VACTERLX|ZNF203|HTX1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZIC3
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human