Order Entry
Denmark
ContactUsLinkComponent
Anti-NEK4 Mouse Monoclonal Antibody [clone: 2E9]
Anti-NEK4 Mouse Monoclonal Antibody [clone: 2E9]
Catalog # ABNOH00006787-M02
Supplier:  Abnova
undefined
Anti-NEK4 Mouse Monoclonal Antibody [clone: 2E9]
Catalog # ABNOH00006787-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NIMA-related kinase 4
  • Antigen symbol:
    NEK4
  • Clonality:
    Monoclonal
  • Clone:
    2E9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    6787
  • Antigen synonyms:
    STK2|NimA-related protein kinase 4|Serine/threonine protein kinase 2|Serine/threonine-protein kinase Nek4|Serine/threonine-protein kinase NRK2|NRK2|pp12301
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: NEK4
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human