Anti-PSMD13 Mouse Polyclonal Antibody
ABNOH00005719-B02P
: Abnova
- Antibody type:Primary
- Antigen name:Proteasome (prosome macropain) 26S Subunit non-ATPase 13
- Antigen symbol:PSMD13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:5719
- Antigen synonyms:Rpn9|S11|HSPC027|p40.5
- Storage buffer:In 1x PBS, pH 7,4
- Sequence:MKDVPGFLQQSQSSGPGQPAVWHRLEELYTKKLWHQLTLQVLDFVQDPCFAQGDGLIKLYENFISEFEHRVNPLSLVEIILHVVRQMTDPNVALTFLEKTREKVKSSDEAVILCKTAIGALKLNIGDLQVTKETIEDVEEMLNNLPGVTSVHSRFYDLSSKYYQTIGNHASYYKDALRFLGCVDIKDLPVSEQQERAFTLGLAGLLGEGVFNFGELLMHPVLESLRNTDRQWLIDTLYAFNSGNVERFQTLKTAWGQQPDLAANEAQLLRKIQLLCLMEMTFTRPANHRQLTFEEIAKSAKITVNEVELLVMKALSVGLVKGSIDEVDKRVHMTWVQPRVLDLQQIKGMKDRLEFWCTDVKSMEMLVEHQAHDILT
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:PSMD13 (NP_002808, 1 a.a. ~ 376 a.a) full-length human protein.
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PSMD13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human