Anti-MAP2K3 Mouse Polyclonal Antibody
ABNOH00005606-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase Kinase 3
- Antigen symbol:MAP2K3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5606
- Antigen synonyms:dual specificity mitogen-activated protein kinase kinase 3|SAPKK-2|MEK 3|MAP kinase kinase 3|MAPKK3|SAPK kinase 2|SAPKK2|PRKMK3|stress-activated protein kinase kinase 2|MKK3|MAPKK 3|MAPK/ERK kinase 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: MAP2K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human