Anti-SRGN Mouse Monoclonal Antibody [clone: 1D8]
ABNOH00005552-M03
: Abnova
- Antibody type:Primary
- Antigen name:Serglycin
- Antigen symbol:SRGN
- Clonality:Monoclonal
- Clone:1D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5552
- Antigen synonyms:PRG1|PRG|PPG
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SRGN
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human