Anti-PLEC Mouse Monoclonal Antibody [clone: 4C3]
Catalog # ABNOH00005339-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Plectin
- Antigen symbol:PLEC
- Clonality:Monoclonal
- Clone:4C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5339
- Antigen synonyms:PLEC1b|EBSOG|EBS1|EBSMD|EBSND|EBSPA|LGMD2Q|PLEC1|PLTN|HD1|PCN|EBSO
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PLEC
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human