Order Entry
Denmark
ContactUsLinkComponent
 
Anti-PCK1 Rabbit Polyclonal Antibody
  ABNOH00005105-D01P
 :  Abnova
undefined
Anti-PCK1 Rabbit Polyclonal Antibody
  ABNOH00005105-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Phosphoenolpyruvate carboxykinase 1
  • Antigen symbol:
    PCK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    5105
  • Antigen synonyms:
    PEPCK1|phosphoenolpyruvate carboxylase|cytosolic phosphoenolpyruvate carboxykinase 1|PCK1|HGNC:8724|phosphopyruvate carboxylase|PEPCKC|PEP carboxykinase|phosphoenolpyruvate carboxykinase 1 (soluble)|PEPCK-C|MGC22652
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: PCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human