Anti-PAK2 Rabbit Polyclonal Antibody
Catalog # ABNOH00005062-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:p21 protein (Cdc42/Rac)-activated kinase 2
- Antigen symbol:PAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5062
- Antigen synonyms:S6/H4 kinase|Serine/threonine-protein kinase PAK 2|Gamma-PAK|p21-activated kinase 2|PAK65
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human