Anti-LMNA Mouse Polyclonal Antibody
Catalog # ABNOH00004000-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lamin A/C
- Antigen symbol:LMNA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4000
- Antigen synonyms:LMNL1|FPLD|FPL|IDC|PRO1|LMNC|CMT2B1|LGMD1B|LMN1|CDCD1|70 kDa lamin|Prelamin-A/C|Lamin-A/C|LFP|FPLD2|LDP1|Renal carcinoma antigen NY-REN-32|CDDC|CMD1A|EMD2|HGPS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: LMNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human