Anti-IMP5 Mouse Monoclonal Antibody [clone: 1C4]
Catalog # ABNOH00003843-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Importin 5
- Antigen symbol:IMP5
- Clonality:Monoclonal
- Clone:1C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3843
- Antigen synonyms:imp5|RANBP5|IMB3|KPNB3|Pse1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IMP5
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human