Anti-ILF2 Mouse Polyclonal Antibody
ABNOH00003608-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Interleukin enhancer binding Factor 2 45kDa
- Antigen symbol:ILF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3608
- Antigen synonyms:MGC8391|PRO3063 |nuclear factor of activated T-cells|NF45|45kDa |interleukin enhancer binding factor 2|45-kDa|ILF2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ILF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human