Anti-GRP Mouse Monoclonal Antibody [clone: 3A11]
ABNOH00002922-M03
: Abnova
- Antibody type:Primary
- Antigen name:Gastrin-releasing Peptide
- Antigen symbol:GRP
- Clonality:Monoclonal
- Clone:3A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:2922
- Antigen synonyms:preproGRP|GRP-10|proGRP|BN
- Storage buffer:In 1x PBS, pH 7,4
- Sequence:MRGRELPLVLLALVLCLAPRGRAVPLPAGGGTVLTKMYPRGNHWAVGHLMGKKSTGESSSVSERGSLKQQLREYIRWEEAARNLLGLIEAKENRNHQPPQPKALGNQQPSWDSEDSSNFKDVGSKGKVGRLSAPGSQREGRNPQLNQQ
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:GRP (AAH04488, 1 a.a. ~ 148 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against a full-length recombinant GRP.
Recommended Dilutions: Western Blot (1 to 5 µg/ml).
Type: Primary
Antigen: GRP
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human