Order Entry
Denmark
ContactUsLinkComponent
Anti-GABBR1 Mouse Monoclonal Antibody [clone: 2D7]
Anti-GABBR1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ABNOH00002550-M01
Supplier:  Abnova
undefined
Anti-GABBR1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ABNOH00002550-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Gamma-Aminobutyric Acid (GABA) B Receptor, 1
  • Antigen symbol:
    GABBR1
  • Clonality:
    Monoclonal
  • Clone:
    2D7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    2550
  • Antigen synonyms:
    GABABR1|GABBR1-3|GPRC3A|GB1|dJ271M21.1.2|dJ271M21.1.1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: GABBR1
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human