Anti-FKBP5 Rabbit Polyclonal Antibody
ABNOH00002289-D01P
: Abnova
- Antibody type:Primary
- Antigen name:FK506 binding protein 5
- Antigen symbol:FKBP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2289
- Antigen synonyms:AIG6|FKBP51|P54|FKBP54|PPIase|Ptg-10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: FKBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human