Anti-F8 Mouse Monoclonal Antibody [clone: 1E8]
ABNOH00002157-M03
: Abnova
- Antibody type:Primary
- Antigen name:Coagulation Factor 8
- Antigen symbol:F8
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2157
- Antigen synonyms:coagulation factor VIII|FVIII|DXS1253E|F8C|RP11-115M6.7|HEMA,antihemophilic factor|factor VIII F8B|AHF|coagulation factor VIIIc|F8B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human