Anti-CRK Mouse Polyclonal Antibody
Catalog # ABNOH00001398-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:V-Crk Avian Sarcoma Virus Ct10 Oncogene Homolog
- Antigen symbol:CRK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1398
- Antigen synonyms:proto-oncogene c-Crk|CRKII|p38|adapter molecule crk
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human