Anti-CNP Rabbit Polyclonal Antibody
ABNOH00001267-D01P
: Abnova
- Antibody type:Primary
- Antigen name:23'-cyclic nucleotide 3' phosphodiesterase
- Antigen symbol:CNP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1267
- Antigen synonyms:CNP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat