Anti-CIDEA Mouse Monoclonal Antibody [clone: 4B9]
Catalog # ABNOH00001149-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cell death-inducing DFFA-like effector a
- Antigen symbol:CIDEA
- Clonality:Monoclonal
- Clone:4B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1149
- Antigen synonyms:Cell death activator CIDE-A|CIDE-A|Cell death-inducing DFFA-like effector A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CIDEA
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human