Anti-BCL3 Rabbit Polyclonal Antibody
Catalog # ABNOH00000602-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:B-cell lymphoma 3-encoded protein
- Antigen symbol:BCL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:602
- Antigen synonyms:Proto-oncogene BCL3|BCL-3|D19S37|B-cell lymphoma 3 protein|BCL3|BCL4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human