Anti-ADRA1A Mouse Monoclonal Antibody [clone: 4C7]
ABNOH00000148-M01
: Abnova
- Antibody type:Primary
- Antigen name:Adrenoceptor Alpha 1A
- Antigen symbol:ADRA1A
- Clonality:Monoclonal
- Clone:4C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:148
- Antigen synonyms:ADRA1L1|ALPHA1AAR|ADRA1C
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ADRA1A
Clonality: Monoclonal
Clone: 4C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human