Order Entry
Denmark
ContactUsLinkComponent
 
Anti-UBOX5 Mouse Monoclonal Antibody [clone: OTI8D5]
  ORIGTA502809
 :  OriGene
undefined
Anti-UBOX5 Mouse Monoclonal Antibody [clone: OTI8D5]
  ORIGTA502809
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    U-box domain containing 5
  • Antigen symbol:
    UBOX5
  • Clonality:
    Monoclonal
  • Clone:
    OTI8D5
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    1 to 130 and 419 to 487
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: UBOX5 (U-box domain containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human